Rat TNF alpha (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Rat
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:The ED50 as determined by the cytolysis of murine L929 cells in the presence of Actinomycin is 0.05ng/ml, corresponding to Specific Activity of 20,000,000 IU/mg.
- Protein synonyms:APC1
- Protein/peptide name:TNF alpha
- Purity:> 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized Tumor Necrosis Factor-alpha in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:The sequence of the first five N-terminal amino acids was determined and was found to be Met-Leu-Arg-Ser-Ser.
- Formulation:The concentrated protein solution (1mg/ml) was lyophilized from 20 mM phosphate buffer and 0.1M NaCl.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together