Order Entry
Italy
ContactUsLinkComponent
Human PTH (1-84) (from E. coli)
Human PTH (1-84) (from E. coli)
Supplier:  Biorbyt
Certificates
Human PTH (1-84) (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The activity calculated by UMR106 cell/cAMP method corresponding to specific activity of 9000 Units/mg.
  • Protein synonyms:
    Parathyrin|PTH1 receptor|PTHY_protein.|PTH1|Parathyroid hormone 1|Parathormone|hPTH|PTHR1|PTHR|PTH|Parathyroid hormone|PTH1R
  • Protein/peptide name:
    PTH (1-84)
  • Purity:
    > 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Parathormone in sterile H2O not less than 100 ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    SVSEIQLMHN LGKHLNSMER VEWLRKKLQD VHNFVALGAP LAPRDAGSQR PRKKEDNVLV ESHEKSLGEA DKADVNVLTK AKSQ.
  • Formulation:
    Each 1.3 mg of lyophilized protein contains 50ug Mannitol and 20 mM PB, pH 7.0.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together