Human Neurotrophin 4 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:Determined by the dose-dependent induction of choline acetyl transferase activity in rat basal forebrain primary septal cell cultures was found to be in the range of 20-50 ng/ml.
- Protein synonyms:NT4
- Protein/peptide name:Neurotrophin 4
- Purity:> 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized NT-4 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:GVSETAPASR RGELAVCDAV SGWVTDRRTA VDLRGREVEV LGEVPAAGGS PLRQYFFETR CKADNAEEGG PGAGGGGCRG VDRRHWVSEC KAKQSYVRAL TADAQGRVGW RWIRIDTACV CTLLSRTGRA.
- Formulation:Lyophilized from concentrated (1mg/ml) solution in water containing 20 mM phosphate buffer pH-7.4 and 150mM NaCl.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together