Order Entry
Italy
ContactUsLinkComponent
Human recombinant MIP4 (from E. coli)
Human recombinant MIP4 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant MIP4 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The Activity is calculated by the ability to chemoattract Human lymphocytes at 1.0-10.0 ng/ml.
  • Protein synonyms:
    Small inducible cytokine A18
  • Protein/peptide name:
    MIP4
  • Purity:
    > 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Macrophage Inflammatory Protein in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Ala-Gln-Val-Gly-Thr.
  • Formulation:
    Lyophilized from 0.2 um filtered concentrated (1.0mg/ml) solution in 20 mM PB, pH 7.4, 100mM NaCl.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together