Order Entry
Italy
ContactUsLinkComponent
 
Human recombinant CXCL9 (from E. coli)
 :  Biorbyt
 
Human recombinant CXCL9 (from E. coli)
 :  Biorbyt

 

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    Determined by its ability to chemoattract human peripheral blood T-Lymphocytes using concentration range of 10.0-100.0 ng/ml.
  • Protein synonyms:
    MIG-1
  • Protein/peptide name:
    CXCL9
  • Purity:
    > 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized MIG in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    TPVVRKGRCS CISTNQGTIH LQSLKDLKQF APSPSCEKIE IIATLKNGVQ TCLNPDSADV KELIKKWEKQ VSQKKKQKNG KKHQKKKVLK VRKSQRSRQK KTT.
  • Formulation:
    Lyophilized from 0.2 um filtered concentrated (1.0mg/ml) solution in 20 mM PB, pH 7.4, 50mM NaCl.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

 

 

Frequently Bought Together