Mouse IL6 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Mouse
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:The ED50 as determined by the dose-dependant induction of 7TD1 cell proliferation, is 0.018 ng/ml.
- Protein synonyms:Interleukin BSF 2
- Protein/peptide name:IL6
- Purity:> 95.0% as determined by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized Interleukin in 10mM HCl to 0.1-1.0 mg/ml to regain full activity, and can then be further diluted to other aqueous solutions.
- Sequence:The sequence of the first five N-terminal amino acids was determined and was found to be Met-Phe-Pro-Thr-Ser.
- Formulation:The protein was lyophilized without any additives.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together