Order Entry
Italy
ContactUsLinkComponent
Human recombinant EPO Fc (from CHO cells)
Human recombinant EPO Fc (from CHO cells)
Supplier:  Biorbyt
Certificates
Human recombinant EPO Fc (from CHO cells)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The ED50 as determined by the dose-dependant stimulation of human megakaryoblastic leukemia cells is less than 2.0 ng/ml, corresponding to Specific Activity of 500,000 Units/mg.
  • Protein synonyms:
    EPO-a
  • Protein/peptide name:
    EPO Fc
  • Purity:
    > 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Erythropoietin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Formulation:
    Each mg of lyophilized powder contains 1x PBS pH-7.4.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together