Order Entry
Italy
ContactUsLinkComponent
 
Human Eotaxin 2 (from E. coli)
 :  Biorbyt
 
Human Eotaxin 2 (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The activity is determined by the chemoattract of human PBE (peripheral blood eosinophils) at concentration between 50-100 ng/ml.
  • Protein synonyms:
    Small-inducible cytokine A24
  • Protein/peptide name:
    Eotaxin 2
  • Purity:
    > 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized CCL24 Human Recombinant in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    VVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKGGVIFT TKKGQQFCGD PKQEWV QRY MKNLDAKQKK ASPRARAVA.
  • Formulation:
    The CCL24 protein was lyophilized from concentrated (1mg/ml) sterile solution containing 20 mM PBS pH-7.4 and 0.15M sodium chloride.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together