Rat EGF (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Rat
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:The ED50 as calculated by the dose-dependant proliferation of murine BALB/c 3T3 cells is less than 0.1ng/ml, corresponding to specific activity of 10,000,000 units/mg.
- Protein synonyms:Urogastrone
- Protein/peptide name:EGF
- Purity:> 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized Rat EGF in sterile water not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:NSNTGCPPSY DGYCLNGGVC MYVESVDRYV CNCVIGYIGE RCQHRDLRWW KLR.
- Formulation:Lyophilized from 0.2 um filtered concentrated (1.0mg/ml) solution in PBS, pH 7.4.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together