Mouse EGF (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Mouse
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:The ED50, calculated by the dose-dependant proliferation of murine BALB/c 3T3 cells (measured by 3H-thymidine uptake) is 0.1 ng/ml.
- Protein synonyms:Urogastrone
- Protein/peptide name:EGF
- Purity:> 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized Epidermal Growth Factor in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Molecular weight:6 kDa
- Sequence:The sequence of the first five N-terminal amino acids was determined and was found to be Asn-Ser-Tyr-Pro-Gly, which agrees with the sequence of native Epidermal Growth Factor human.
- Formulation:The protein was lyophilized from concentrated (1mg/ml) solution containing 1x PBS pH-7.4.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together