Order Entry
Italy
ContactUsLinkComponent
 
Human BMP7 (from E. coli)
 :  Biorbyt
 
Human BMP7 (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    BMP 7
  • Protein/peptide name:
    BMP7
  • Purity:
    > 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Bone Morphogenetic Protein-7 in sterile 20 mM-100mM AcOH (acetic Acid) not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Ser-Thr-Gly-Ser-Lys.
  • Formulation:
    BMP-7 was lyophilized from concentrated (1mg/ml) sterile solution containing 10mM sodium citrate pH=3.5.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together