Order Entry
Italy
ContactUsLinkComponent
Avidin (from egg white)
Avidin (from egg white)
Supplier:  Biorbyt
Certificates
Avidin (from egg white)
Supplier:  Biorbyt

Specifications

  • Source:
    Egg white
  • Storage conditions:
    Avidin although stable at 4 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Please prevent freeze-thaw cycles.
  • Biological activity:
    Greater than 11 U/mg, unit binds 1ug biotin.
  • Protein synonyms:
    AVD
  • Protein/peptide name:
    Avidin
  • Solubility:
    It is recommended to reconstitute the lyophilized Avidin in sterile H2O not less than 100ug/ml or more than 10mg/ml solutions.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together