Human recombinant PI3KD946GST (from insect cells (Sf9))
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Sorgente luminosa:Insect cells (Sf9)
- Specie:Human
- Condizioni di conservazione:PI3Kb although stable at 4 °C for week, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Please prevent freeze-thaw cycles.
- Protein synonyms:Phosphoinositide 3-kinase p110d
- Protein/peptide name:PI3KD946GST
- Purezza:Greater than 95.0% as determined by SDS Page.
- Formulazione:0.5mg/ml solution in 25mM Tris, pH 8.0, 50mM NaCl, 0.5mM MgCl2, and 50% glycerol.
- Applicazioni testate:SDS-PAGE
Specifications
Frequently Bought Together