Human recombinant APO GST (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.
- Protein synonyms:Apolipoprotein D
- Protein/peptide name:APO GST
- Formulation:APO-D protein at 100ug/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.
- Tested applications:SDS-PAGE
Frequently Bought Together