Mouse recombinant b NGF
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Species:Mouse
- Storage conditions:Store at −20 °C.
- Biological activity:The ED50 as determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 106 units/mg.
- UniProtKB:P01139
- Protein/peptide name:b NGF
- Purity:>98%
- Solubility:It is recommended to reconstitute the lyophilized Murine NGF-beta in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:SSTHPVFHMG EFSVCDSVSV WVGDKTTATD IKGKEVTVLA EVNINNSVFR QYFFETKCRA SNPVESGCRG IDSKHWNSYC TTTHTFVKAL TTDEKQAAWR FIRIDTACVC VLSRKATRRG
- Formulation:The Recombinant Mouse b-NGF was lyophilized without additives.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together