Order Entry
Italy
ContactUsLinkComponent
Mouse recombinant IL7
Mouse recombinant IL7
Supplier:  Biorbyt
Certificates
Mouse recombinant IL7
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Storage conditions:
    Store at −20 °C.
  • Biological activity:
    The ED50 as determined by the dose-dependent stimulation of the proliferation of Mouse 2E8 cells is ≤ 0.2 ng/ml, corresponding to a specific activity of ≥ 5 x 106 units/mg.
  • Protein synonyms:
    Lymphopoietin 1|LP-1
  • UniProtKB:
    P10168
  • Protein/peptide name:
    IL7
  • Purity:
    >98%
  • Solubility:
    It is recommended to reconstitute the lyophilized Interleukin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    MECHIKDKEG KAYESVLMIS IDELDKMTGT DSNCPNNEPN FFRKHVCDDT KEAAFLNRAA RKLKQFLKMN ISEEFNVHLL TVSQGTQTLV NCTSKEEKNV KEQKKNDACF LKRLLREIKT CWNKILKGSI
  • Formulation:
    Lyophilized from concentrated (1mg/ml) solution in water containing no additives.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together