Order Entry
Italy
ContactUsLinkComponent
Rat stem cell factor
Rat stem cell factor
Supplier:  Biorbyt
Certificates
Rat stem cell factor
Supplier:  Biorbyt

Specifications

  • Species:
    Rat
  • Storage conditions:
    Store at −20 °C.
  • Biological activity:
    Determined by the dose-dependent stimulation of the proliferation of the human TF-1 cells. The expected ED50 for this effect is 20 - 40 ng/ml.
  • Protein synonyms:
    Kit ligand Precursor
  • UniProtKB:
    P21581
  • Protein/peptide name:
    Stem Cell Factor
  • Purity:
    >98%
  • Solubility:
    It is recommended to reconstitute the lyophilized SCF in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    MQEICRNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV THLSVSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVAC MEENAPKNVK ESLKKPETRN FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA
  • Formulation:
    Lyophilized from concentrated (1mg/ml) solution in water containing 0.02% NaHCO3.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together