Rat recombinant prolactin
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Species:Rat
- Storage conditions:Store at −20 °C.
- Biological activity:Determined by its ability to induce the proliferation of rat Nb2-11 cells in the concentration range of 0.1-1.0 ng/ml.
- Gene ID:24683
- Protein synonyms:Mammotropin
- UniProtKB:P01237
- Protein/peptide name:Prolactin
- Purity:>98%
- Solubility:It is recommended to reconstitute the lyophilized Prolactin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:MLPVCSGGDC QTPLPELFDR VVMLSHYIHT LYTDMFIEFD KQYVQDREFI AKAINDCPTS SLATPEDKEQ AQKVPPEVLL NLILSLVHSW NDPLFQLITG LGGIHEAPDA IISRAKEIEE QNKRLLEGIE KIISQAYPEA KGNEIYLVWS QLPSLQGVDE ESKDLAFYNN IRCLRRDSHK VDNYLKFLRC QIVHKNNC
- Formulation:The protein was lyophilized from concentrated (1mg/ml) solution with 10mM sodium Phosphate buffer pH=8 and 50mM Nacl.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together