Order Entry
Italy
ContactUsLinkComponent
Human recombinant Artemin
Human recombinant Artemin
Supplier:  Biorbyt
Certificates
Human recombinant Artemin
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C.
  • Biological activity:
    Determined by it's ability to promote survival and neurite outgrowth and dorsal root ganglion neurons.
  • Protein synonyms:
    ARTN|Enovin|Neublastin NBN|EVN|ART
  • UniProtKB:
    Q5T4W7
  • Protein/peptide name:
    Artemin
  • Purity:
    >98%
  • Solubility:
    It is recommended to reconstitute the lyophilized Artemin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    AGGPGSRARA AGARGCRLRS QLVPVRALGL GHRSDELVRF RFCSGSCRRA RSPHDLSLAS LLGAGALRPP PGSRPVSQPC CRPTRYEAVS FMDVNSTWRT VDRLSATACG CLG
  • Formulation:
    Artemin was lyophilized after extensive dialysis against 10mM sodium citrate pH-4.5 and 150mM sodium chloride.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together