Order Entry
Italy
ContactUsLinkComponent
 
Human BDNF
 :  Biorbyt
 
Human BDNF
 :  Biorbyt

 

  • Species:
    Human
  • Storage conditions:
    Store at −20 °C.
  • Biological activity:
    Determined by its ability to stimulate the proliferation of rat C6 cells. The expected ED50 for this effect is 0.5-1.0 ug/ml.
  • Protein synonyms:
    Abrineurin
  • UniProtKB:
    P23560
  • Protein/peptide name:
    BDNF
  • Purity:
    >98%
  • Solubility:
    It is recommended to reconstitute the lyophilized Brain-derived Neurotrophic Factor in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    MHSDPARRGE LSVCDSISEW VTAADKKTAV DMSGGTVTVL EKVPVSKGQL KQYFYETKCN PMGYTKEGCR GIDKRHWNSQ CRTTQSYVRA LTMDSKKRIG WRFIRIDTSC VCTLTIKRGR
  • Formulation:
    The protein was lyophilized after dialysis against 20 mM sodium citrate buffer pH=5.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

 

 

Frequently Bought Together