Order Entry
Italy
ContactUsLinkComponent
Mouse recombinant CXCL9
Mouse recombinant CXCL9
Supplier:  Biorbyt
Certificates
Mouse recombinant CXCL9
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Storage conditions:
    Store at −20 °C.
  • Biological activity:
    Determined by its ability to chemoacttract human lymphocytes using a concentration range of 0.1-1.0 ng/ml.
  • Protein synonyms:
    MIG-1
  • UniProtKB:
    P18340
  • Protein/peptide name:
    CXCL9
  • Purity:
    >98%
  • Solubility:
    It is recommended to reconstitute the lyophilized MIG in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    TLVIRNARCS CISTSRGTIH YKSLKDLKQF APSPNCNKTE IIATLKNGDQ TCLDPDSANV KKLMKEWEKK INQKKKQKRG KKHQKNMKNR KPKTPQSRRR SRKTT
  • Formulation:
    The CXCL9 was lyophilized from concentrated (1mg/ml) solution in water containing no additives.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together