Order Entry
Italy
Orders LinkContactUsLinkComponent
Disposal Bags
Disposal Bags
Supplier:   Brand
Disposal Bags
Supplier:  Brand
Certificates
Disposal Bags
2 Options Available Below
Product Details & Documents
Bags: Disposal bag, PP/PA.
Holders:
Epoxy-coated steel wire, with rubber feet. Non-autoclavable.
Epoxy-coated steel wire, with rubber feet. Non-autoclavable.
Caution: risk of injury. Never put sharp objects such as needles into plastic bags. Bags should stay open during sterilisation.
Documents
Product Family Options
Bags
Product Information
  • Material
    L×W
    Availability
    Price
  • 129-0131
    Sterilisation/disposal bags, autoclavable (121 °C)
    Sterilisation/disposal bags, autoclavable (121 °C)
    PP
    300×200 mm
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 19,70 €
    19,70 €
    1 * 100 pezzi
       19,70 €
  • 129-9812
    Sterilisation/disposal bags, autoclavable (134 °C) and hot-air sterilisation at 180 °C
    Sterilisation/disposal bags, autoclavable (134 °C) and hot-air sterilisation at 180 °C
    PA
    300×200 mm
    Log in for availability
    Log in for availability
    1 * 50 pezzi
    1 * 50 pezzi - 34,30 €
    34,30 €
    1 * 50 pezzi
       34,30 €
Holders
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Stand for disposal bags, white
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 10,60 €
    10,60 €
    1 * 1 pezzi
       10,60 €