Order Entry
Italy
ContactUsLinkComponent
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)
Catalog # 1B1509-25G
Supplier:  VWR Chemicals
CAS Number:  
undefined
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)
Catalog # 1B1509-25G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
CAS Number:9007-49-2
 :73049-39-5
MDL Number:MFCD00130921