Order Entry
Italy
ContactUsLinkComponent
QuEChERS extraction salt packets Q-sep™
  REST23992
 :  Restek
undefined
QuEChERS extraction salt packets Q-sep™
  REST23992
 :  Restek
  • Description:
    Q-sep QuEChERS extraction salt packets (original unbuffered methods) with 4 g magnesium sulphate and 1,0 g sodium chloride-original unbuffered
  • Seq:
    03
  • Pk:
    50
Q-sep™ QuEChERS provides fast, simple sample preparation for multi residue pesticide analysis.
  • Preweighed, ultra-pure sorbents
  • Convenient and easy to use
  • Salt packets eliminate the need for a second empty tube to transfer salts
Support original unbuffered, AOAC (2007.01) and European (EN 15662) QuEChERS methods.