Order Entry
Italy
ContactUsLinkComponent
Anti-KRT8 Mouse Monoclonal Antibody [clone: C-43]
Anti-KRT8 Mouse Monoclonal Antibody [clone: C-43]
  ABNOMAB14320
 :  Abnova
Anti-KRT8 Mouse Monoclonal Antibody [clone: C-43]
  ABNOMAB14320
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Keratin 8
  • Antigen symbol:
    KRT8
  • Clonality:
    Monoclonal
  • Clone:
    C-43
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    3856
  • Amino acid number:
    P05787
  • Storage buffer:
    In 10 mM PBS.
  • Immunogen:
    Cytoskeletal preparation from HeLa cells.
  • Purification:
    Protein A/G purification
  • Size:
    100 µG
  • Pk:
    100 µG

 

 

Mouse monoclonal antibody raised against native human KRT8.

Type: Primary
Antigen: KRT8
Clonality: Monoclonal
Clone: C-43
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human