Anti-Cdkn1b Mouse Monoclonal Antibody [clone: DCS-72.F6]
ABNOMAB14203
: Abnova
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase inhibitor 1B
- Antigen symbol:CDKN1B
- Clonality:Monoclonal
- Clone:DCS-72.F6
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Form:Liquid
- Gene ID:12576
- Amino acid number:P46414
- Storage buffer:In PBS.
- Immunogen:Recombinant protein corresponding to full length mouse Cdkn1b.
- Purification:Protein G purification
- Size:100 µG
- Pk:100 µG
Mouse monoclonal antibody raised against full length recombinant mouse Cdkn1b.
Type: Primary
Antigen: Cdkn1b
Clonality: Monoclonal
Clone: DCS-72.F6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse