Anti-SYT5 Rabbit Polyclonal Antibody
Catalog # ABGEAP14064C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Synaptotagmin 5
- Antigen symbol:SYT5
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:Syt5|SytV|Synaptotagmin V|Synaptotagmin 5|Synaptotagmin IX
- Size:100 µg
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SYT5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human