Anti-ADAM8 Rabbit Polyclonal Antibody
Catalog # ABGEAP14058A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase Domain 8
- Antigen symbol:ADAM8
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:CD156a|CD156|MS2
- Size:100 µg
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ADAM8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human