Order Entry
Italy
ContactUsLinkComponent
Anti-KRT76 Mouse Monoclonal Antibody (CF488A) [clone: KRTH/1076]
Anti-KRT76 Mouse Monoclonal Antibody (CF488A) [clone: KRTH/1076]
Supplier:  Biotium
Certificates
Anti-KRT76 Mouse Monoclonal Antibody (CF488A) [clone: KRTH/1076]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC881076-500
  • BNC881076-100
  • Antigen symbol:
  • KRT76
  • KRT76
  • Antigen name:
  • Cytokeratin HMW
  • Cytokeratin HMW
  • Antigen synonyms:
  • Keratin|Type-II Keratin Kb9|Cytokeratin-2P|KRT2P|Cytokeratin 2|HUMCYT2A|K2P|CK-2P|Type II Cytoskeletal 2 Oral|K76|Keratin 2p|Keratin-76|KRT2B
  • Keratin|Type-II Keratin Kb9|Cytokeratin-2P|KRT2P|Cytokeratin 2|HUMCYT2A|K2P|CK-2P|Type II Cytoskeletal 2 Oral|K76|Keratin 2p|Keratin-76|KRT2B
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone KRTH/1076
  • Clone KRTH/1076
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 654392
  • 654392
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together