Anti-CD48 Mouse Monoclonal Antibody (CF640R) [clone: 5-4.8]
Supplier: Biotium
Certificates
About this item
Reacts with human CD48, a 45 kDa glycosyl phophatidyl-inositol (GPI)-anchored cell surface protein. CD48 is strongly expressed on lymphocytes and monocytes and weakly on granulocytes but is absent on platelets, fibroblasts, epithelium and endothelium. CD48 is one of the markers for detecting the defects of GPI anchoring structure on the patients with paroxysmal nocturnal hemoglobulinuria (PNH) and serves as a low affinity ligand for CD2.
Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: 5-4.8
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC401057-100
- BNC401057-500
- Antigen symbol:
- CD48
- CD48
- Antigen synonyms:
- BCM1 surface antigen|B cell membrane protein|SLAM family member 2 (SLAMF2)|BLAST1|Signaling lymphocytic activation molecule 2|TCT.1|B-cell activation marker
- BCM1 surface antigen|B cell membrane protein|SLAM family member 2 (SLAMF2)|BLAST1|Signaling lymphocytic activation molecule 2|TCT.1|B-cell activation marker
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone 5-4.8
- Clone 5-4.8
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 243564
- 243564
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together