Order Entry
Italy
ContactUsLinkComponent
 
Anti-LGR5 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNLS1235G
Supplier:  Novus Biologicals
Anti-LGR5 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNLS1235G
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Leucine-rich Repeat Containing G protein-coupled receptor 5
  • Antigen symbol:
    LGR5
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Gene ID:
    8549
  • Antigen synonyms:
    GPR67|GPR49|G-protein coupled receptor 49|LGR5|G-protein coupled receptor HG38|G-protein coupled receptor 67|Leucine-rich repeat-containing G-protein coupled receptor 5
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: LGR5
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human