About this item
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. This antibody cocktail recognizes acidic (Type I or LMW) and basic (Type II or HMW) cytokeratins, with 67 kDa (CK1); 64 kDa (CK3); 59 kDa (CK4); 58 kDa (CK5); 56 kDa (CK6); 55 kDa (CK7); 52 kDa (CK8); 56.5 kDa (CK10); 53 kDa (CK13); 50 kDa (CK14); 50 kDa (CK15); 48 kDa (CK16); 46 kDa (CK17); 45 kDa (CK18) and 40 kDa (CK19). Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. KRT-PAN is a broad spectrum anti pan-cytokeratin antibody cocktail, which differentiates epithelial tumors from non-epithelial tumors e.g. squamous vs. adenocarcinoma of the lung, liver carcinoma, breast cancer, and esophageal cancer. It is useful in characterizing the source of various neoplasms and to study the distribution of cytokeratin containing cells in epithelia during normal development and during the development of epithelial neoplasms. This antibody stains cytokeratins present in normal and abnormal human tissues and shows high sensitivity in the recognition of epithelial cells and carcinomas.
Type: Primary
Antigen: KRT
Clonality: Monoclonal
Clone: KRT7/760 / KRT17/ 778 / KRT18/834 / KRTL/828 / KRTH/829 / KRT/457
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Chicken, Cow, Dog, Human, Monkey, Mouse, Rabbit, Rat
Specifications
- Catalog No:
- BNC470999-500
- BNC470999-100
- Antigen symbol:
- KRT
- KRT
- Antigen name:
- Cytokeratin
- Cytokeratin
- Antigen synonyms:
- K77|Keratin|KRT1B|Type-II Keratin Kb39|Keratin-77|type II cytoskeletal 1b|K1B|CK-1B|Keratin 1B|Cytokeratin-1B
- K77|Keratin|KRT1B|Type-II Keratin Kb39|Keratin-77|type II cytoskeletal 1b|K1B|CK-1B|Keratin 1B|Cytokeratin-1B
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- Clone KRT7/760 / KRT17/ 778 / KRT18/834 / KRTL/828 / KRTH/829 / KRT/457
- Clone KRT7/760 / KRT17/ 778 / KRT18/834 / KRTL/828 / KRTH/829 / KRT/457
- Reactivity:
- Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
- Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 334989 (KRT77) , 654392 (KRT76)
- 334989 (KRT77) , 654392 (KRT76)
- Isotype:
- IgGs
- IgGs
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide