Order Entry
Italy
ContactUsLinkComponent
 
Anti-SIAH1/SIAH2 Mouse Monoclonal Antibody [clone: 8G7H12]
 
undefined
Anti-SIAH1/SIAH2 Mouse Monoclonal Antibody [clone: 8G7H12]

 

  • Catalog No:
  • NOVUNB110-93596
  • NOVUNB110-93596SS
  • Antigen symbol:
  • SIAH1/SIAH2
  • SIAH1/SIAH2
  • Antigen name:
  • siah E3 ubiquitin protein ligase 1
  • siah E3 ubiquitin protein ligase 1
  • Antigen synonyms:
  • Seven in Absentia Homolog 1/2
  • Seven in Absentia Homolog 1/2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 8G7H12
  • 8G7H12
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 6477
  • 6477
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together