Anti-RAD51L1 Mouse Monoclonal Antibody [clone: 1 H3/13]
Supplier: Novus Biologicals
Certificates
About this item
Mouse Monoclonal Rad51L1 Antibody (1 H3/13). Tested Applications: Western Blot. Tested Reactivity: Human, Primate, Mouse (-).
Type: Primary
Antigen: RAD51L1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- NOVUNB100-176SS
- NOVUNB100-176
- Antigen symbol:
- RAD51L1
- RAD51L1
- Antigen name:
- RAD51-like 1
- RAD51-like 1
- Antigen synonyms:
- REC2|RAD51L1|R51H2
- REC2|RAD51L1|R51H2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 1 H3/13
- 1 H3/13
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 5890
- 5890
- Isotype:
- IgG2a
- IgG2a
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together