Order Entry
Italy
ContactUsLinkComponent
 
Anti-Hepatocyte Specific Mouse Monoclonal Antibody [clone: HepPar1]
 :  Biotium
 
Anti-Hepatocyte Specific Mouse Monoclonal Antibody [clone: HepPar1]
 :  Biotium

 

  • Catalog No:
  • BNUB0966-100
  • BNUB0966-500
  • Antigen name:
  • Hepatocyte Specific
  • Hepatocyte Specific
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone HepPar1
  • Clone HepPar1
  • Reactivity:
  • Human, Dog
  • Human, Dog
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

 

Frequently Bought Together