Anti-MMP2 Mouse Monoclonal Antibody [clone: MMP2/8B4] (HRP (Horseradish Peroxidase))
Catalog # NOVUNB200-114H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Matrix Metalloproteinase 2
- Antigen symbol:MMP2
- Clonality:Monoclonal
- Clone:MMP2/8B4
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:4313
- Antigen synonyms:MMP-2|72 kDa gelatinase|matrix metalloproteinase-2|collagenase type IV-A|neutrophil gelatinase|gelatinase A|matrix metalloproteinase-II|72 kDa type IV collagenase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Mouse Monoclonal MMP2 Antibody (MMP2/8B4) [HRP]. Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human, Mouse, Rat.
Recommended Dilutions: Immunohistochemistry (Parafin): 2 ?g/ml
Type: Primary
Antigen: MMP2
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human