Order Entry
Italy
ContactUsLinkComponent
Anti-MUC5AC Mouse Monoclonal Antibody (CF405S) [clone: MUC5AC/917]
Anti-MUC5AC Mouse Monoclonal Antibody (CF405S) [clone: MUC5AC/917]
Supplier:  Biotium
Certificates
Anti-MUC5AC Mouse Monoclonal Antibody (CF405S) [clone: MUC5AC/917]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC040917-100
  • BNC040917-500
  • Antigen symbol:
  • MUC5AC
  • MUC5AC
  • Antigen name:
  • Mucin 5AC
  • Mucin 5AC
  • Antigen synonyms:
  • Mucin 5AC oligomeric mucus/gel forming|Mucin 5 Subtypes A And C|Apomucin Major Airway Glycoprotein|Tracheobronchial Mucin (TBM)|Mucin 5 subtype AC tracheobronchial
  • Mucin 5AC oligomeric mucus/gel forming|Mucin 5 Subtypes A And C|Apomucin Major Airway Glycoprotein|Tracheobronchial Mucin (TBM)|Mucin 5 subtype AC tracheobronchial
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone MUC5AC/917
  • Clone MUC5AC/917
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 534332
  • 534332
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together