Anti-CDKN1B Mouse Monoclonal Antibody (Purified, BSA-free) [clone: DCS-72.F6]
Catalog # BTIUBNUM0771-50
Supplier: Biotium
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase inhibitor 1B
- Antigen symbol:CDKN1B
- Clonality:Monoclonal
- Clone:Clone DCS-72.F6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Gene ID:238990
- Antigen synonyms:CDKN1B|CDKN4|KIP1|Cyclin-dependent kinase inhibitor p27 Kip1|MEN1B|Cyclin Dependent Kinase Inhibitor 1B|MEN4
- Storage buffer:PBS/0,05% azide
- Concentration:1 mg/mL
- Purification:BSA-free
- Size:50 μL
- Pk:50 µl
Specifications
About this item
Recognizes a 27 kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. Its epitope spans between aa 83-204 of p27. It is highly specific and shows no cross-reaction with other related mitotic inhibitors. p27Kip1 functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF- induced G1 arrest. p27Kip1 is a candidate tumor suppressor gene. This MAb co-precipitates cdk4 in complex p27Kip1 and is excellent for staining of formalin-fixed tissues.
Type: Primary
Antigen: CDKN1B
Clonality: Monoclonal
Clone: DCS-72.F6
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Mouse, Rat