Order Entry
Italy
ContactUsLinkComponent
Anti-CD5 Mouse Monoclonal Antibody (CF594) [clone: C5/473 CD5/54/F6]
Anti-CD5 Mouse Monoclonal Antibody (CF594) [clone: C5/473 CD5/54/F6]
Supplier:  Biotium
Certificates
Anti-CD5 Mouse Monoclonal Antibody (CF594) [clone: C5/473 CD5/54/F6]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC940748-500
  • BNC940748-100
  • Antigen symbol:
  • CD5
  • CD5
  • Antigen synonyms:
  • CD5 antigen (p56 62)|Ly12|LEU1|Lymphocyte glycoprotein T1/Leu1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|LyA
  • CD5 antigen (p56 62)|Ly12|LEU1|Lymphocyte glycoprotein T1/Leu1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|LyA
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone C5/473 CD5/54/F6
  • Clone C5/473 CD5/54/F6
  • Reactivity:
  • Rabbit, Human, Bovine, Rat
  • Rabbit, Human, Bovine, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 58685
  • 58685
  • Isotype:
  • IgGs
  • IgGs
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together