Anti-TRAP2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-9363R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:TNFR-associated protein 2
- Antigen symbol:TRAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:5708
- Antigen synonyms:TRAP2|Rpn1|P97|55.11 protein|26S proteasome non-ATPase regulatory subunit 2|non-ATPase|proteasome (prosome|26S proteasome regulatory subunit S2|S2|macropain) 26S subunit|proteasome 26S non-ATPase subunit 2|TNFR-associated protein 2|PSMD2|2|MGC14274|tumor necrosis factor receptor-associated protein 2|26S proteasome subunit p97
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:525-575/908
- Purification:Purified by Protein A
- Size:100 μL
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: TRAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat