Order Entry
Italy
ContactUsLinkComponent
Anti-KRT8 Mouse Monoclonal Antibody (CF647) [clone: C-43]
Anti-KRT8 Mouse Monoclonal Antibody (CF647) [clone: C-43]
Supplier:  Biotium
Certificates
Anti-KRT8 Mouse Monoclonal Antibody (CF647) [clone: C-43]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC470688-500
  • BNC470688-100
  • Antigen symbol:
  • KRT8
  • KRT8
  • Antigen name:
  • Cytokeratin 8
  • Cytokeratin 8
  • Antigen synonyms:
  • Keratin 8|CYK8|Cytokeratin Endo A|Type-II Keratin Kb8|KRT8|K8|CARD2|DreK8|K2C8|Krt 2.8|CYKER|EndoA|CK8
  • Keratin 8|CYK8|Cytokeratin Endo A|Type-II Keratin Kb8|KRT8|K8|CARD2|DreK8|K2C8|Krt 2.8|CYKER|EndoA|CK8
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone C-43
  • Clone C-43
  • Reactivity:
  • Pig, Rabbit, Sheep, Human, Bovine
  • Pig, Rabbit, Sheep, Human, Bovine
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 533782 , 708445
  • 533782 , 708445
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together