Anti-CTCF Rabbit Polyclonal Antibody
100 μL
Catalog # BOSSBS-3778R
Supplier: Bioss
Jump to:
Product Details & Documents
Type: Primary
Antigen: CTCF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
bs-3778R
Datasheet
Specifications
- Size:100 μL
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Purified by Protein A
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Store at –20 °C for 12 months
- Host:Rabbit
- Concentration:1 ug/ul
- Cross adsorption:No
- Shipping temperature:4 °C
- Antigen name:CCCTC-binding Factor
- Cat. No.:BOSSBS-3778R
- Reactivity:Human|Rat|Mouse
- Gene ID:10664
- Antigen synonyms:transcriptional repressor CTCF|CTCFL paralog|11-zinc finger protein|11 zinc finger transcriptional repressor
- Immunogen:470-520/727
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:CTCF
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.



