Order Entry
Italy
ContactUsLinkComponent
 
Anti-CCNB1 Mouse Monoclonal Antibody (CF405S) [clone: V92.1]
 :  Biotium
 
Anti-CCNB1 Mouse Monoclonal Antibody (CF405S) [clone: V92.1]
 :  Biotium

 

  • Catalog No:
  • BNC040680-500
  • BNC040680-100
  • Antigen symbol:
  • CCNB1
  • CCNB1
  • Antigen name:
  • Cyclin B1
  • Cyclin B1
  • Antigen synonyms:
  • CCNB1_HUMAN|CCNB1|CCNB|G2 Mitotic Specific Cyclin B1
  • CCNB1_HUMAN|CCNB1|CCNB|G2 Mitotic Specific Cyclin B1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone V92.1
  • Clone V92.1
  • Reactivity:
  • Hamster, Human, Mouse
  • Hamster, Human, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 23960
  • 23960
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together