Order Entry
Italy
ContactUsLinkComponent
Anti-KRT77 & KRT76 Mouse Monoclonal Antibody (CF568) [clone: AE-1 / AE-3]
Anti-KRT77 & KRT76 Mouse Monoclonal Antibody (CF568) [clone: AE-1 / AE-3]
Supplier:  Biotium
Certificates
Anti-KRT77 & KRT76 Mouse Monoclonal Antibody (CF568) [clone: AE-1 / AE-3]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC680371-100
  • BNC680371-500
  • Antigen symbol:
  • KRT77 & KRT76
  • KRT77 & KRT76
  • Antigen name:
  • Cytokeratin
  • Cytokeratin
  • Antigen synonyms:
  • K77|Keratin|KRT1B|Type-II Keratin Kb39|Keratin-77|type II cytoskeletal 1b|K1B|CK-1B|Keratin 1B|Cytokeratin-1B
  • K77|Keratin|KRT1B|Type-II Keratin Kb39|Keratin-77|type II cytoskeletal 1b|K1B|CK-1B|Keratin 1B|Cytokeratin-1B
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone AE-1 / AE-3
  • Clone AE-1 / AE-3
  • Reactivity:
  • Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
  • Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 334989 (KRT77) , 654392 (KRT76)
  • 334989 (KRT77) , 654392 (KRT76)
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together