Order Entry
Italy
ContactUsLinkComponent
Anti-AFP Mouse Monoclonal Antibody (CF405S) [clone: C3]
Anti-AFP Mouse Monoclonal Antibody (CF405S) [clone: C3]
Supplier:  Biotium
Certificates
Anti-AFP Mouse Monoclonal Antibody (CF405S) [clone: C3]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC040354-500
  • BNC040354-100
  • Antigen symbol:
  • AFP
  • AFP
  • Antigen name:
  • AFP
  • AFP
  • Antigen synonyms:
  • FETA|Alpha fetoglobulin|HPAFP
  • FETA|Alpha fetoglobulin|HPAFP
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone C3
  • Clone C3
  • Reactivity:
  • Pig, Monkey, Human, Dog
  • Pig, Monkey, Human, Dog
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 518808
  • 518808
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together