Anti-SLC3A2 Mouse Monoclonal Antibody (Purified) [clone: IPO-T10]
: Biotium
CD98 exits as a heterodimer containing a disulphide-linked glycosylated heavy chain and a non-glycosylated light chain. It is a member of the solute carrier family and encodes a cell surface, transmembrane protein. The protein exists as the heavy chain of a heterodimer, covalently bound through disulfide bonds to one of several possible light chains. The encoded transporter plays a role in regulation of intracellular calcium levels and transports L-type amino acids. Alternatively spliced transcript variants, encoding different isoforms, have been characterized.
Type: Primary
Antigen: SLC3A2
Clonality: Monoclonal
Clone: IPO-T10
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
- Catalog No:
- BNUB0337-500
- BNUB0337-100
- Antigen symbol:
- Solute carrier family 3, member 2
- Solute carrier family 3, member 2
- Antigen name:
- Solute carrier family 3 member 2
- Solute carrier family 3 member 2
- Antigen synonyms:
- 4F2HC|Slc3a2|E16|4F2|NACAE|CD98HC|hLAT1|MDU1|LAT1|MPE16|Solute Carrier Family 3 (activators of dibasic and neutral amino acid transport) Member 2|4T2HC
- 4F2HC|Slc3a2|E16|4F2|NACAE|CD98HC|hLAT1|MDU1|LAT1|MPE16|Solute Carrier Family 3 (activators of dibasic and neutral amino acid transport) Member 2|4T2HC
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Clone IPO-T10
- Clone IPO-T10
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 502769
- 502769
- Isotype:
- IgM
- IgM
- ImmunoFluorescence:
- Yes
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage buffer:
- PBS/0,05% BSA/0,05% azide
- PBS/0,05% BSA/0,05% azide
Frequently Bought Together