Order Entry
Italy
ContactUsLinkComponent
Anti-TAG-72 / CA72.4 Mouse Monoclonal Antibody (CF568) [clone: B72.3]
Anti-TAG-72 / CA72.4 Mouse Monoclonal Antibody (CF568) [clone: B72.3]
Supplier:  Biotium
Certificates
Anti-TAG-72 / CA72.4 Mouse Monoclonal Antibody (CF568) [clone: B72.3]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC680276-100
  • BNC680276-500
  • Antigen name:
  • TAG-72 / CA72.4
  • TAG-72 / CA72.4
  • Antigen synonyms:
  • CA72.4|Tumor associated glycoprotein 72
  • CA72.4|Tumor associated glycoprotein 72
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone B72.3
  • Clone B72.3
  • Reactivity:
  • Hamster, Human, Bovine, Dog, Rat
  • Hamster, Human, Bovine, Dog, Rat
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together