Anti-CD5 Mouse Monoclonal Antibody (CF640R) [clone: Cris-1]
Supplier: Biotium
Certificates
About this item
Recognizes a 67 kDa transmembrane protein, which is identified as CD5 (HLDA I; WS Code T 29HLDA III; WS Code T 530). The CD5 antigen is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. Anti-CD5 is a pan T-cell marker that also reacts with a range of neoplastic B-cells, e.g. chronic lymphocytic leukemia/small lymphocytic lymphoma (CLL/SLL), mantle cell lymphoma, and a subset (~10%) of diffuse large B-cell lymphoma. CD5 aberrant expression is useful in making a diagnosis of mature T-cell neoplasms.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: Cris-1
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Rhesus monkey
Specifications
- Catalog No:
- BNC400176-100
- BNC400176-500
- Simbolo dell'antigene:
- CD5
- CD5
- Sinomini antigene:
- CD5 antigen (p56 62)|Ly12|LEU1|Lymphocyte glycoprotein T1/Leu1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|LyA
- CD5 antigen (p56 62)|Ly12|LEU1|Lymphocyte glycoprotein T1/Leu1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|LyA
- Tipo anticorpo:
- Primary
- Primary
- Clonalità:
- Monoclonale
- Monoclonale
- Coniugazione:
- CF® 640R
- CF® 640R
- Clone:
- Clone Cris-1
- Clone Cris-1
- Reattività:
- Rhesus monkey, Human
- Rhesus monkey, Human
- Ospite:
- Mouse
- Mouse
- ID gene:
- 58685
- 58685
- Isotipo:
- IgG2a kappa
- IgG2a kappa
- ImmunoFluorescenza:
- Yes
- Yes
- Concentrazione:
- 0,1 mg/ml
- 0,1 mg/ml
- Tampone di conservazione:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together