Anti-TAG-72 / CA72.4 Mouse Monoclonal Antibody [clone: CA72/145]
Catalog # BTIUBNUM0145-50
Supplier: Biotium
Specifications
- Antibody type:Primary
- Antigen name:TAG-72 / CA72.4
- Clonality:Monoclonal
- Clone:Clone CA72/145
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:CA72.4|Tumor associated glycoprotein 72
- Storage buffer:PBS/0,05% azide
- Concentration:1 mg/mL
- Purification:BSA-free
- Size:50 μL
- Pk:50 µl
Specifications
About this item
Recognizes an oncofetal antigen of 220 kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin. The majority of human adenocarcinomas including colorectal, pancreatic, gastric, ovarian, endometrial, mammary, and non-small cell lung cancer display some cell populations that are positive for TAG72. About 60% of carcinoma patients express TAG72 in their sera. TAG72 is reportedly useful in distinguishing pulmonary adenocarcinomas from pleural mesotheliomas.
- Purified, BSA-free
Type: Primary
Antigen: TAG-72 / CA72.4
Clonality: Monoclonal
Clone: CA72/145
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human