Order Entry
Italy
ContactUsLinkComponent
Anti-TAG-72 / CA72.4 Mouse Monoclonal Antibody [clone: CA72/145]
Anti-TAG-72 / CA72.4 Mouse Monoclonal Antibody [clone: CA72/145]
Supplier:  Biotium
Certificates
Anti-TAG-72 / CA72.4 Mouse Monoclonal Antibody [clone: CA72/145]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB0145-100
  • BNUB0145-500
  • Antigen name:
  • TAG-72 / CA72.4
  • TAG-72 / CA72.4
  • Antigen synonyms:
  • CA72.4|Tumor associated glycoprotein 72
  • CA72.4|Tumor associated glycoprotein 72
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone CA72/145
  • Clone CA72/145
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Frequently Bought Together